Search
Search

GSTZ1| Gene Symbol: GSTZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493361.4 | XP_001493411.1 | 646 | 94 | ||
| XM_014735741.2 | XP_014591227.1 | 1887 | 94 | |||
| XM_023628183.1 | XP_023483951.1 | 1591 | 94 | |||
| XM_023628184.1 | XP_023483952.1 | 599 | 94 | |||
Set Membership: |
|