Search
Search
| Gene Symbol: MPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632080.1 | XP_023487848.1 | 1653 | 70 | ||
| XM_023632081.1 | XP_023487849.1 | 2007 | 70 | |||
| XM_023632082.1 | XP_023487850.1 | 1807 | 70 | |||
| XM_023632083.1 | XP_023487851.1 | 1853 | 70 | |||
| XM_023632084.1 | XP_023487852.1 | 1650 | 70 | |||
| XM_023632086.1 | XP_023487854.1 | 1635 | 70 | |||
| XM_023632087.1 | XP_023487855.1 | 1632 | 70 | |||
| XM_023632088.1 | XP_023487856.1 | 1616 | 70 | |||
| XM_023632089.1 | XP_023487857.1 | 1598 | 70 | |||
| XM_023632090.1 | XP_023487858.1 | 1559 | 70 | |||
Set Membership: |
|