Search
Search

HSD17B2| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738466.2 | XP_014593952.2 | 580 | 111 | ||
| XM_023637383.1 | XP_023493151.1 | 579 | 111 | |||
| XM_023637384.1 | XP_023493152.1 | 715 | 111 | |||
Set Membership: |
|