Search
Search
| Gene Symbol: DOK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638455.1 | XP_023494223.1 | 661 | 50 | ||
| XM_023638456.1 | XP_023494224.1 | 639 | 50 | |||
| XM_023638457.1 | XP_023494225.1 | 545 | 50 | |||
| XM_023638458.1 | XP_023494226.1 | 763 | 50 | |||
Set Membership: |
|