Search
Search

PLEKHG1| Gene Symbol: PLEKHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738207.2 | XP_014593693.2 | 3788 | 81 | ||
| XM_023632920.1 | XP_023488688.1 | 4954 | 81 | |||
| XM_023632921.1 | XP_023488689.1 | 3521 | 81 | |||
| XM_023632922.1 | XP_023488690.1 | 3415 | 81 | |||
| XM_023632923.1 | XP_023488691.1 | 3402 | 81 | |||
| XM_023632924.1 | XP_023488692.1 | 5309 | 81 | |||
| XM_023632925.1 | XP_023488693.1 | 3408 | 81 | |||
Set Membership: |
|