Search
Search
| Gene Symbol: GSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638825.1 | XP_023494593.1 | 317 | 104 | ||
| XM_023638826.1 | XP_023494594.1 | 317 | 104 | |||
| XM_023638827.1 | XP_023494595.1 | 533 | 104 | |||
| XM_023638828.1 | XP_023494596.1 | 317 | 104 | |||
| XM_023638829.1 | XP_023494597.1 | 317 | 104 | |||
| XM_023638830.1 | XP_023494598.1 | 317 | 104 | |||
Set Membership: |
|