Search
Search

VPS50| Gene Symbol: VPS50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492642.5 | XP_001492692.1 | 2328 | 71 | ||
| XM_023639201.1 | XP_023494969.1 | 2347 | 71 | |||
| XM_023639202.1 | XP_023494970.1 | 2369 | 71 | |||
| XM_023639203.1 | XP_023494971.1 | 2683 | 71 | |||
| XM_023639204.1 | XP_023494972.1 | 2185 | 71 | |||
| GenBank mRNA | JL637761.1 | - | 2 - 3 | 228 | 71 | |
Set Membership: |
|