Search
Search

RNF34| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612609.3 | XP_005612666.1 | 812 | 111 | ||
| XM_005612610.3 | XP_005612667.1 | 757 | 111 | |||
| XM_023647200.1 | XP_023502968.1 | 776 | 111 | |||
Set Membership: |
|