Search
Search

CFAP251| Gene Symbol: CFAP251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647206.1 | XP_023502974.1 | 1191 | 64 | ||
| XM_023647207.1 | XP_023502975.1 | 1277 | 64 | |||
| XM_023647208.1 | XP_023502976.1 | 1059 | 64 | |||
| XM_023647209.1 | XP_023502977.1 | 1191 | 64 | |||
Set Membership: |
|