Search
Search
| Gene Symbol: PUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647354.1 | XP_023503122.1 | 1427 | 74 | ||
| XM_023647355.1 | XP_023503123.1 | 1181 | 74 | |||
| XM_023647356.1 | XP_023503124.1 | 1240 | 74 | |||
Set Membership: |
|