Search
Search

MALT1| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647884.1 | XP_023503652.1 | 1697 | 79 | ||
| XM_023647885.1 | XP_023503653.1 | 1664 | 79 | |||
| XM_023647887.1 | XP_023503655.1 | 1698 | 79 | |||
| GenBank mRNA | JL626500.1 | - | 7 - 8 | 792 | 79 | |
Set Membership: |
|