Search
Search

ZNF236| Gene Symbol: ZNF236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612986.3 | XP_005613043.1 | 3544 | 76 | ||
| XM_023648000.1 | XP_023503768.1 | 3735 | 76 | |||
| XM_023648001.1 | XP_023503769.1 | 3735 | 76 | |||
| XM_023648002.1 | XP_023503770.1 | 3648 | 76 | |||
| XM_023648003.1 | XP_023503771.1 | 3633 | 76 | |||
| XM_023648004.1 | XP_023503772.1 | 3735 | 76 | |||
| XM_023648005.1 | XP_023503773.1 | 3710 | 76 | |||
| XM_023648006.1 | XP_023503774.1 | 3886 | 76 | |||
| XM_023648007.1 | XP_023503775.1 | 3648 | 76 | |||
| XM_023648008.1 | XP_023503776.1 | 3908 | 76 | |||
| XM_023648009.1 | XP_023503777.1 | 3738 | 76 | |||
| XM_023648010.1 | XP_023503778.1 | 3735 | 76 | |||
Set Membership: |
|