Search
Search

LYPLA1| Gene Symbol: LYPLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613164.3 | XP_005613221.3 | 123 | 100 | ||
| XM_023648610.1 | XP_023504378.1 | 402 | 100 | |||
| XM_023648611.1 | XP_023504379.1 | 403 | 100 | |||
| XM_023648613.1 | XP_023504381.1 | 375 | 100 | |||
Set Membership: |
|