Search
Search
| Gene Symbol: ZFAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613297.3 | XP_005613354.1 | 731 | 76 | ||
| XM_014728094.2 | XP_014583580.1 | 719 | 76 | |||
| XM_014728095.2 | XP_014583581.1 | 722 | 76 | |||
| XM_023648996.1 | XP_023504764.1 | 722 | 76 | |||
| XM_023648997.1 | XP_023504765.1 | 1262 | 76 | |||
| XM_023648998.1 | XP_023504766.1 | 467 | 76 | |||
| XM_023648999.1 | XP_023504767.1 | 729 | 76 | |||
| XM_023649000.1 | XP_023504768.1 | 730 | 76 | |||
Set Membership: |
|