Search
Search
| Gene Symbol: OFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917181.5 | XP_001917216.3 | 1649 | 89 | ||
| XM_023633905.1 | XP_023489673.1 | 1823 | 89 | |||
| XM_023633906.1 | XP_023489674.1 | 1848 | 89 | |||
| XM_023633907.1 | XP_023489675.1 | 1721 | 89 | |||
| XM_023633908.1 | XP_023489676.1 | 1823 | 89 | |||
| XM_023633909.1 | XP_023489677.1 | 1971 | 89 | |||