Search
Search

PPEF1| Gene Symbol: PPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728774.2 | XP_014584260.2 | 1798 | 79 | ||
| XM_023634450.1 | XP_023490218.1 | 1589 | 79 | |||
| XM_023634451.1 | XP_023490219.1 | 1715 | 79 | |||
| XM_023634452.1 | XP_023490220.1 | 1726 | 79 | |||
| XM_023634453.1 | XP_023490221.1 | 1488 | 79 | |||
| XM_023634454.1 | XP_023490222.1 | 1697 | 79 | |||
| XM_023634455.1 | XP_023490223.1 | 1440 | 79 | |||
| XM_023634456.1 | XP_023490224.1 | 1491 | 79 | |||
| XM_023634457.1 | XP_023490225.1 | 1628 | 79 | |||
| XM_023634458.1 | XP_023490226.1 | 1401 | 79 | |||
| XM_023634459.1 | XP_023490227.1 | 367 | 79 | |||
| XM_023634460.1 | XP_023490228.1 | 1339 | 79 | |||
| XM_023634461.1 | XP_023490229.1 | 1390 | 79 | |||
| XM_023634462.1 | XP_023490230.1 | 367 | 79 | |||
| XM_023634463.1 | XP_023490231.1 | 264 | 79 | |||
Set Membership: |
|