Search
Search

HUWE1| Gene Symbol: HUWE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728912.2 | XP_014584398.2 | 3886 | 94 | ||
| XM_023634500.1 | XP_023490268.1 | 3886 | 94 | |||
| XM_023634501.1 | XP_023490269.1 | 3876 | 94 | |||
| XM_023634502.1 | XP_023490270.1 | 3769 | 94 | |||
| XM_023634503.1 | XP_023490271.1 | 3870 | 94 | |||
| XM_023634504.1 | XP_023490272.1 | 3792 | 94 | |||
| XM_023634505.1 | XP_023490273.1 | 3859 | 94 | |||
| XM_023634507.1 | XP_023490275.1 | 3886 | 94 | |||
| XM_023634508.1 | XP_023490276.1 | 3886 | 94 | |||
| XM_023634509.1 | XP_023490277.1 | 3886 | 94 | |||
Set Membership: |
|