Search
Search
| Gene Symbol: ADAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918220.4 | XP_001918255.1 | 859 | 99 | ||
| XM_005603081.3 | XP_005603138.1 | 859 | 99 | |||
| XM_005603082.3 | XP_005603139.1 | 859 | 99 | |||
| XM_005603083.3 | XP_005603140.1 | 859 | 99 | |||
| GenBank mRNA | JL619781.1 | - | 3 - 4 | 298 | 99 | |
Set Membership: |
|