Search
Search

KIFAP3| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014739518.2 | XP_014595004.2 | 806 | 157 | ||
| XM_023640528.1 | XP_023496296.1 | 805 | 157 | |||
| XM_023640529.1 | XP_023496297.1 | 513 | 157 | |||
| XM_023640530.1 | XP_023496298.1 | 807 | 157 | |||
| XM_023640531.1 | XP_023496299.1 | 601 | 157 | |||
| XM_023640532.1 | XP_023496300.1 | 698 | 157 | |||
Set Membership: |
|