Search
Search

GATAD2B| Gene Symbol: GATAD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014739900.2 | XP_014595386.2 | 1012 | 113 | ||
| XM_023641114.1 | XP_023496882.1 | 1011 | 113 | |||
| XM_023641115.1 | XP_023496883.1 | 727 | 113 | |||
| XM_023641116.1 | XP_023496884.1 | 714 | 113 | |||
Set Membership: |
|