Search
Search

RBM20| Gene Symbol: RBM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003363408.4 | XP_003363456.1 | 1674 | 64 | ||
| XM_023639469.1 | XP_023495237.1 | 1357 | 64 | |||
| XM_023639475.1 | XP_023495243.1 | 1306 | 64 | |||
| XM_023639481.1 | XP_023495249.1 | 1393 | 64 | |||
| XM_023639485.1 | XP_023495253.1 | 1397 | 64 | |||
| XM_023639492.1 | XP_023495260.1 | 1336 | 64 | |||
Set Membership: |
|