Search
Search
| Gene Symbol: MTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001914916.5 | XP_001914951.2 | 2244 | 64 | ||
| XM_023646964.1 | XP_023502732.1 | 2668 | 64 | |||
| XM_023646970.1 | XP_023502738.1 | 842 | 64 | |||
Set Membership: |
|