Search
Search

CD276| Gene Symbol: CD276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654011.1 | XP_023509779.1 | 1155 | 77 | ||
| XM_023654016.1 | XP_023509784.1 | 1423 | 77 | |||
| XM_023654020.1 | XP_023509788.1 | 1186 | 77 | |||
| XM_023654027.1 | XP_023509795.1 | 1042 | 77 | |||
Set Membership: |
|