Search
Search

ARIH1| Gene Symbol: ARIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654140.1 | XP_023509908.1 | 1022 | 122 | ||
| XM_023654149.1 | XP_023509917.1 | 1022 | 122 | |||
| XM_023654154.1 | XP_023509922.1 | 811 | 122 | |||
Set Membership: |
|