Search
Search

GALK2| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499722.5 | XP_001499772.4 | 1135 | 67 | ||
| XM_014733659.2 | XP_014589145.2 | 1103 | 67 | |||
| XM_023617694.1 | XP_023473462.1 | 1174 | 67 | |||
| XM_023617705.1 | XP_023473473.1 | 1075 | 67 | |||
| XM_023617713.1 | XP_023473481.1 | 923 | 67 | |||
Set Membership: |
|