Search
Search

FXYD1| Gene Symbol: FXYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362228.4 | XP_003362276.1 | 385 | 78 | ||
| XM_005596145.1 | XP_005596202.1 | 371 | 78 | |||
| XM_005596146.3 | XP_005596203.1 | 365 | 78 | |||
| XM_023649537.1 | XP_023505305.1 | 464 | 78 | |||
Set Membership: |
|