Search
Search

SHKBP1| Gene Symbol: SHKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362315.3 | XP_003362363.2 | 154 | 123 | ||
| XM_023649768.1 | XP_023505536.1 | 154 | 123 | |||
| XM_023649769.1 | XP_023505537.1 | 154 | 123 | |||
Set Membership: |
|