Search
Search
| Gene Symbol: TMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651918.1 | XP_023507686.1 | 641 | 82 | ||
| XM_023651919.1 | XP_023507687.1 | 588 | 82 | |||
| XM_023651920.1 | XP_023507688.1 | 935 | 82 | |||
| XM_023651921.1 | XP_023507689.1 | 871 | 82 | |||
| XM_023651922.1 | XP_023507690.1 | 525 | 82 | |||
| XM_023651923.1 | XP_023507691.1 | 503 | 82 | |||
| XM_023651924.1 | XP_023507692.1 | 886 | 82 | |||
| XM_023651925.1 | XP_023507693.1 | 1153 | 82 | |||
| XM_023651926.1 | XP_023507694.1 | 609 | 82 | |||
| XM_023651927.1 | XP_023507695.1 | 639 | 82 | |||
| XM_023651928.1 | XP_023507696.1 | 642 | 82 | |||
| XM_023651930.1 | XP_023507698.1 | 645 | 82 | |||
Set Membership: |
|