Search
Search

SPATA22| Gene Symbol: SPATA22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504720.3 | XP_001504770.1 | 882 | 158 | ||
| XM_023653268.1 | XP_023509036.1 | 1156 | 158 | |||
| XM_023653269.1 | XP_023509037.1 | 885 | 158 | |||
Set Membership: |
|