Search
Search
| Gene Symbol: ACCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729433.2 | XP_014584919.1 | 1769 | 64 | ||
| XM_023653908.1 | XP_023509676.1 | 1059 | 64 | |||
| XM_023653909.1 | XP_023509677.1 | 1059 | 64 | |||
| XM_023653910.1 | XP_023509678.1 | 768 | 64 | |||
| XM_023653911.1 | XP_023509679.1 | 1757 | 64 | |||
| XM_023653912.1 | XP_023509680.1 | 943 | 64 | |||
| XM_023653913.1 | XP_023509681.1 | 856 | 64 | |||
Set Membership: |
|