Search
Search

C13H16orf96| Gene Symbol: C13H16orf96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616398.1 | XP_023472166.1 | 3926 | 58 | ||
| XM_023616399.1 | XP_023472167.1 | 3119 | 58 | |||
| XM_023616400.1 | XP_023472168.1 | 3924 | 58 | |||
| XM_023616401.1 | XP_023472169.1 | 3906 | 58 | |||
| XM_023616402.1 | XP_023472170.1 | 3937 | 58 | |||
| XM_023616403.1 | XP_023472171.1 | 3926 | 58 | |||
Set Membership: |
|