Search
Search

DNAJC18| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504229.4 | XP_001504279.1 | 756 | 75 | ||
| XM_023617604.1 | XP_023473372.1 | 1102 | 75 | |||
| GenBank mRNA | JL622515.1 | - | 3 - 4 | 395 | 75 | |
Set Membership: |
|