Search
Search

EFCAB13| Gene Symbol: EFCAB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652311.1 | XP_023508079.1 | 3370 | 114 | ||
| XM_023652313.1 | XP_023508081.1 | 2386 | 114 | |||
| XM_023652314.1 | XP_023508082.1 | 2553 | 114 | |||
| XM_023652315.1 | XP_023508083.1 | 3371 | 114 | |||
| XM_023652316.1 | XP_023508084.1 | 3296 | 114 | |||
| XM_023652317.1 | XP_023508085.1 | 3370 | 114 | |||
| XM_023652318.1 | XP_023508086.1 | 1945 | 114 | |||
| XM_023652319.1 | XP_023508087.1 | 3371 | 114 | |||
| XM_023652320.1 | XP_023508088.1 | 3532 | 114 | |||
Set Membership: |
|