Search
Search

HYKK| Gene Symbol: HYKK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277459.1 | NP_001264388.1 | 2 - 3 | 381 | 69 | |
| XM_015278879.2 | XP_015134365.2 | 2 - 3 | 832 | 69 | ||
| XM_015278880.2 | XP_015134366.2 | 2 - 3 | 840 | 69 | ||
| XM_015278881.2 | XP_015134367.1 | 2 - 3 | 455 | 69 | ||
| XM_015278882.2 | XP_015134368.1 | 2 - 3 | 477 | 69 | ||
| XM_015278883.2 | XP_015134369.1 | 2 - 3 | 364 | 69 | ||
| XM_025153828.1 | XP_025009596.1 | 2 - 3 | 2198 | 69 | ||
| GenBank mRNA | BX932491.1 | - | 2 - 3 | 414 | 69 | |
| BX933588.2 | - | 2 - 3 | 381 | 69 | ||
| BX935072.1 | - | 1 - 2 | 474 | 69 | ||
Set Membership: |
|