Search
Search

KIF1B| Gene Symbol: KIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947461.3 | XP_004947518.1 | 2 - 3 | 469 | 92 | |
| XM_015297159.2 | XP_015152645.1 | 2 - 3 | 470 | 92 | ||
| XM_015297160.2 | XP_015152646.1 | 2 - 3 | 470 | 92 | ||
| XM_015297162.2 | XP_015152648.1 | 2 - 3 | 470 | 92 | ||
| XM_015297163.2 | XP_015152649.1 | 2 - 3 | 470 | 92 | ||
| XM_015297165.2 | XP_015152651.1 | 2 - 3 | 469 | 92 | ||
| XM_025142650.1 | XP_024998418.1 | 2 - 3 | 470 | 92 | ||
| XM_025142651.1 | XP_024998419.1 | 2 - 3 | 470 | 92 | ||
| XM_025142652.1 | XP_024998420.1 | 2 - 3 | 470 | 92 | ||
| XM_025142654.1 | XP_024998422.1 | 2 - 3 | 469 | 92 | ||
| XM_417608.6 | XP_417608.4 | 2 - 3 | 469 | 92 | ||
| GenBank mRNA | BX950339.1 | - | 2 - 3 | 248 | 92 | |
Set Membership: |
|