Search
Search

LOC420860| Gene Symbol: LOC420860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256499.1 | NP_001243428.1 | 1 - 2 | 180 | 131 | |
| GenBank mRNA | BX930895.2 | - | 1 - 2 | 180 | 131 | |
| BX930914.2 | - | 1 - 2 | 149 | 131 | ||
| BX931360.2 | - | 1 - 2 | 157 | 131 | ||
Set Membership: |
|