Search
Search

COL26A1| Gene Symbol: COL26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946472.3 | XP_004946529.1 | 2 - 3 | 437 | 68 | |
| XM_015295986.2 | XP_015151472.1 | 2 - 3 | 438 | 68 | ||
| XM_015295988.2 | XP_015151474.1 | 2 - 3 | 438 | 68 | ||
| XM_015295989.2 | XP_015151475.1 | 2 - 3 | 437 | 68 | ||
| XM_015295991.2 | XP_015151477.1 | 2 - 3 | 263 | 68 | ||
| XM_015295993.2 | XP_015151479.1 | 2 - 3 | 289 | 68 | ||
| GenBank mRNA | CR387598.1 | - | 2 - 3 | 214 | 68 | |
Set Membership: |
|