Search
Search

EME1| Gene Symbol: EME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642357.3 | XP_003642405.1 | 7 - 8 | 1646 | 64 | |
| XM_003642358.3 | XP_003642406.1 | 7 - 8 | 1628 | 64 | ||
| XM_004946332.2 | XP_004946389.1 | 7 - 8 | 1706 | 64 | ||
| XM_015295521.1 | XP_015151007.1 | 7 - 8 | 1688 | 64 | ||
| XM_025141851.1 | XP_024997619.1 | 6 - 7 | 946 | 64 | ||
| GenBank mRNA | AB266761.1 | - | 8 - 9 | 1399 | 64 | |
| AB266762.1 | - | 8 - 9 | 1027 | 64 | ||
| AB266763.1 | - | 7 - 8 | 913 | 64 | ||
| AB266765.1 | - | 4 - 5 | 559 | 64 | ||
| CR386766.1 | - | 7 - 8 | 907 | 64 | ||
| CR523212.1 | - | 6 - 7 | 1032 | 64 | ||
Set Membership: |
|