Search
Search

TM4SF1a| Gene Symbol: TM4SF1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277830.1 | NP_001264759.1 | 5 - 6 | 594 | 97 | |
| NM_001277831.1 | NP_001264760.1 | 5 - 6 | 591 | 97 | ||
| NM_001277832.1 | NP_001264761.1 | 4 - 5 | 497 | 97 | ||
| XM_015280003.2 | XP_015135489.1 | 4 - 5 | 1677 | 97 | ||
| GenBank mRNA | BU240401.1 | - | 5 - 6 | 571 | 97 | |
| BU243566.1 | - | 5 - 6 | 97 | |||
| BU246972.1 | - | 1 - 2 | 153 | 97 | ||
| BX932079.2 | - | 4 - 5 | 517 | 97 | ||
| BX935444.1 | - | 5 - 6 | 571 | 97 | ||
Set Membership: |
|