Search
Search

USP40| Gene Symbol: USP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257318.1 | NP_001244247.1 | 23 - 24 | 2794 | 94 | |
| XM_015289068.2 | XP_015144554.1 | 23 - 24 | 3001 | 94 | ||
| XM_015289069.2 | XP_015144555.1 | 24 - 25 | 3104 | 94 | ||
| XM_015289070.2 | XP_015144556.1 | 24 - 25 | 3099 | 94 | ||
| XM_015289071.2 | XP_015144557.1 | 24 - 25 | 3100 | 94 | ||
| XM_015289072.2 | XP_015144558.1 | 22 - 23 | 2836 | 94 | ||
| XM_015289073.2 | XP_015144559.1 | 23 - 24 | 3002 | 94 | ||
| XM_025152062.1 | XP_025007830.1 | 24 - 25 | 3095 | 94 | ||
Set Membership: |
|