Search
Search

SLC51A| Gene Symbol: SLC51A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277697.1 | NP_001264626.1 | 5 - 6 | 607 | 71 | |
| GenBank mRNA | BU372751.1 | - | 1 - 2 | 59 | 71 | |
| BX934769.2 | - | 5 - 6 | 584 | 71 | ||
Set Membership: |
|