Search
Search

PLIN4| Gene Symbol: PLIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317088.1 | NP_001304017.1 | 7 - 8 | 990 | 81 | |
| XM_004948798.2 | XP_004948855.1 | 7 - 8 | 998 | 81 | ||
| XM_015299884.2 | XP_015155370.1 | 7 - 8 | 1101 | 81 | ||
| XM_025144244.1 | XP_025000012.1 | 7 - 8 | 984 | 81 | ||
| GenBank mRNA | BX929460.2 | - | 6 - 7 | 978 | 81 | |
| BX934069.2 | - | 5 - 6 | 813 | 81 | ||
Set Membership: |
|