Search
Search

Rbms2| Gene Symbol: Rbms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861283.3 | XP_004861340.2 | 10 - 11 | 1135 | 56 | |
| XM_004861284.2 | XP_004861341.1 | 10 - 11 | 1135 | 56 | ||
| XM_004861285.3 | XP_004861342.1 | 10 - 11 | 1135 | 56 | ||
| XM_013078269.2 | XP_012933723.1 | 10 - 11 | 1135 | 56 | ||
| XM_021241464.1 | XP_021097123.1 | 12 - 13 | 1267 | 56 | ||
| XM_021241465.1 | XP_021097124.1 | 10 - 11 | 912 | 56 | ||
| XM_021241466.1 | XP_021097125.1 | 11 - 12 | 1170 | 56 | ||
| XM_021241467.1 | XP_021097126.1 | 10 - 11 | 931 | 56 | ||
Set Membership: |
|