| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000587473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
ENST00000587473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014941.hg.1 |
ENST00000252674.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014941.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014941.hg.1 |
TC19001091.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000586154.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000587473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
ENST00000587473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009417.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
NM_005934.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
TC1900009417.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
ENST00000252674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
ENST00000585588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175542.hg.1 |
NM_005934.3 |
Refseq |