| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19018655.hg.1 |
ENST00000251203.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19018655.hg.1 |
ENST00000559264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19018655.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19018655.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19018655.hg.1 |
TC19001304.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000558276.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010067.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
NR_038198.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
TC1900010067.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000251203.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000557978.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000558222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
ENST00000559735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
NM_025245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900182950.hg.1 |
NR_038198.1 |
Refseq |