| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000334731.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000377031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000473252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000479413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
ENST00000486911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007547.hg.1 |
NM_182505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013332.hg.1 |
ENST00000460975.1 |
Ensembl |