| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000342108.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399135.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399136.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000399139.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
ENST00000478313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146078.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_001146079.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_012130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008081.hg.1 |
NM_144492.2 |
Refseq |