Search
Search

TMPRSS13| Gene Symbol: TMPRSS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579777.2 | XP_005579834.1 | 9 - 10 | 4518 | 83 | |
| XM_015435779.1 | XP_015291265.1 | 7 - 8 | 3126 | 83 | ||
Set Membership: |
|