Search
Search

MAP3K13| Gene Symbol: MAP3K13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546588.2 | XP_005546645.1 | 3 - 4 | 2412 | 93 | |
| XM_005546589.2 | XP_005546646.1 | 3 - 4 | 1163 | 93 | ||
| XM_005546590.2 | XP_005546647.1 | 3 - 4 | 2429 | 93 | ||
| XM_005546591.2 | XP_005546648.1 | 3 - 4 | 786 | 93 | ||
| XM_015445615.1 | XP_015301101.1 | 3 - 4 | 2413 | 93 | ||
| XM_015445616.1 | XP_015301102.1 | 3 - 4 | 2412 | 93 | ||
Set Membership: |
|