Search
Search

TMEM269| Gene Symbol: TMEM269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543712.2 | XP_005543769.1 | 4 - 5 | 501 | 89 | |
| XM_015437403.1 | XP_015292889.1 | 4 - 5 | 501 | 89 | ||
Set Membership: |
|